Learn More
ATCC MAVER-1; Mantle Cell Lymphoma; Human (Homo sapiens)
Catalog No. 502382633
Encompass
MAVER-1 is a lymphoblast cell line isolated in 2003 from the peripheral blood of a 77-year-old male with mantle cell lymphoma. This is a hypodiploid cell line with a chromosome number range of 38-40 and can be used for immunology research.
Catalog No. 50-238-2633 Supplier ATCC Supplier No. CRL3008
MAVER-1 is a lymphoblast cell line isolated in 2003 from the peripheral blood of a 77-year-old male with mantle cell lymphoma. This is a hypodiploid cell line with a chromosome number range of 38-40 and can be used for immunology research.