Learn More
Medchemexpress LLC HY-15257A 2mg Medchemexpress, Mavoglurant (racemate) CAS:1636881-61-2 Purity:>98%
SDP
Catalog No. 501932730
Encompass_Preferred
Medchemexpress, HY-15257A 2mg Mavoglurant (racemate) CAS:1636881-61-2 Mavoglurant (racemate) is the racemate of mavoglurant. Mavoglurant is a novel, non-competitive mGlu5 receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2730 Supplier Medchemexpress LLC Supplier No. HY15257A2MG
Medchemexpress, HY-15257A 2mg Mavoglurant (racemate) CAS:1636881-61-2 Mavoglurant (racemate) is the racemate of mavoglurant. Mavoglurant is a novel, non-competitive mGlu5 receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.