Learn More
Cayman Chemical MavorIxafor 10mg
SDP
Catalog No. 502940331
Encompass_Preferred
A CXCR4 antagonist (IC50 = 13 nM in a radioligand binding assay); selective for CXCR4 over CCR1, CCR2b, CCR4, CCR5, CXCR1, and CXCR2 (IC50s = >10 µM for all); reduces CXCL12-induced chemotaxis of CCRF-CEM T cells (IC50 = 19 nM); inhibits fusion of CHO-K1 cells expressing gp120 to P4 MAGI cells IC50 = 1.5 nM; decreases HIV-1 NL4.3 viral replication in MT-4 cells and isolated human PBMCs (IC50s = 2 and 9 nM, respectively)
Catalog No. 50-294-0331 Supplier Cayman Chemical Supplier No. 2843910 MG
A CXCR4 antagonist (IC50 = 13 nM in a radioligand binding assay); selective for CXCR4 over CCR1, CCR2b, CCR4, CCR5, CXCR1, and CXCR2 (IC50s = >10 µM for all); reduces CXCL12-induced chemotaxis of CCRF-CEM T cells (IC50 = 19 nM); inhibits fusion of CHO-K1 cells expressing gp120 to P4 MAGI cells IC50 = 1.5 nM; decreases HIV-1 NL4.3 viral replication in MT-4 cells and isolated human PBMCs (IC50s = 2 and 9 nM, respectively)