Learn More
Medchemexpress LLC Mavorixafor hydrochloride | 880549-30-4 | 99.6% | C21H28ClN5 | 25 MG
SDP
Catalog No. 5000358030
Encompass_Preferred
Mavorixafor hydrochloride is a potent, selective, and orally available CXCR4 antagonist. It exhibits an IC50 value of 13 nM against CXCR4 125I-SDF binding. This compound also inhibits the replication of T-tropic HIV-1 (NL4.3 strain) in MT-4 cells and PBMCs with IC50 values of 1 nM and 9 nM, respectively, making it suitable for WHIM syndrome research. It demonstrates no effect on other chemokine receptors such as CCR1, CCR2b, CCR4, CCR5, CXCR1, and CXCR2.
  • Potent and selective CXCR4 antagonist
  • Orally available for ease of administration
  • Inhibits T-tropic HIV-1 replication
  • Useful for WHIM syndrome studies
  • Suppresses anchorage-dependent cell growth
  • Impairs cell motility and inhibits migration
  • Prevents Matrigel invasion and lung metastasis in in vivo models
Catalog No. 50-003-58030 Supplier Medchemexpress LLC Supplier No. 25MGHY50101C
Mavorixafor hydrochloride is a potent, selective, and orally available CXCR4 antagonist. It exhibits an IC50 value of 13 nM against CXCR4 125I-SDF binding. This compound also inhibits the replication of T-tropic HIV-1 (NL4.3 strain) in MT-4 cells and PBMCs with IC50 values of 1 nM and 9 nM, respectively, making it suitable for WHIM syndrome research. It demonstrates no effect on other chemokine receptors such as CCR1, CCR2b, CCR4, CCR5, CXCR1, and CXCR2.
  • Potent and selective CXCR4 antagonist
  • Orally available for ease of administration
  • Inhibits T-tropic HIV-1 replication
  • Useful for WHIM syndrome studies
  • Suppresses anchorage-dependent cell growth
  • Impairs cell motility and inhibits migration
  • Prevents Matrigel invasion and lung metastasis in in vivo models