Learn More
Medchemexpress LLC Mavorixafor hydrochloride | 880549-30-4 | 99.6% | C21H28ClN5 | 50 MG
SDP
Catalog No. 5000358106
Encompass_Preferred
Mavorixafor hydrochloride is a potent and selective CXCR4 antagonist. It inhibits the replication of T-tropic HIV-1 in MT-4 cells and PBMCs. This compound can be used for the study of WHIM syndrome and has shown no effect on other chemokine receptors.
  • Potent and selective CXCR4 antagonist
  • Inhibits HIV-1 replication in MT-4 cells and PBMCs
  • Suitable for WHIM syndrome research
  • Orally available compound
  • Shows no effect on other tested chemokine receptors
Catalog No. 50-003-58106 Supplier Medchemexpress LLC Supplier No. HY50101C50MG
Mavorixafor hydrochloride is a potent and selective CXCR4 antagonist. It inhibits the replication of T-tropic HIV-1 in MT-4 cells and PBMCs. This compound can be used for the study of WHIM syndrome and has shown no effect on other chemokine receptors.
  • Potent and selective CXCR4 antagonist
  • Inhibits HIV-1 replication in MT-4 cells and PBMCs
  • Suitable for WHIM syndrome research
  • Orally available compound
  • Shows no effect on other tested chemokine receptors