Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mavorixafor hydrochloride | 880549-30-4 | MFCD11977316 | 99.1% | 385.93 g·mol⁻1 | C21H28ClN5 | 10 MG
SDP

Catalog No. 5000298758
Encompass_Preferred

Mavorixafor hydrochloride is a potent, selective CXCR4 antagonist supplied as the hydrochloride salt for research use. It is provided as a viscous liquid with high purity and is intended for in vitro and preclinical studies of CXCR4 signaling and related disease models.

  • High purity: 99.1%.
  • Molecular weight: 385.93 g·mol⁻1.
  • Cas number: 880549-30-4.
  • Physical form: viscous liquid, light yellow to brown.
  • Storage: store at -20 °C, sealed, protect from moisture and light.
  • Application: research into CXCR4 biology and WHIM syndrome models.

Catalog No. 50-002-98758 Supplier Medchemexpress LLC Supplier No. HY50101C10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mavorixafor hydrochloride is a potent, selective CXCR4 antagonist supplied as the hydrochloride salt for research use. It is provided as a viscous liquid with high purity and is intended for in vitro and preclinical studies of CXCR4 signaling and related disease models.

  • High purity: 99.1%.
  • Molecular weight: 385.93 g·mol⁻1.
  • Cas number: 880549-30-4.
  • Physical form: viscous liquid, light yellow to brown.
  • Storage: store at -20 °C, sealed, protect from moisture and light.
  • Application: research into CXCR4 biology and WHIM syndrome models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.