Learn More
Medchemexpress LLC HY-50101A 10mg Medchemexpress, Mavorixafor (trihydrochloride) CAS:2309699-17-8 Purity:>98%
SDP
Catalog No. 502029319
Encompass_Preferred
Medchemexpress, HY-50101A 10mg Mavorixafor (trihydrochloride) CAS:2309699-17-8 Mavorixafor trihydrochloride (AMD-070 trihydrochloride) is a potent, selective and orally available CXCR4 antagonist, with an IC50 value of 13 nM against CXCR4 125I-SDF binding, and also inhibits the replication of T-tropic HIV-1 (NL4.3 strain) in MT-4 cells and PBMCs with an IC50 of 1 and 9 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9319 Supplier Medchemexpress LLC Supplier No. HY50101A10MG
Medchemexpress, HY-50101A 10mg Mavorixafor (trihydrochloride) CAS:2309699-17-8 Mavorixafor trihydrochloride (AMD-070 trihydrochloride) is a potent, selective and orally available CXCR4 antagonist, with an IC50 value of 13 nM against CXCR4 125I-SDF binding, and also inhibits the replication of T-tropic HIV-1 (NL4.3 strain) in MT-4 cells and PBMCs with an IC50 of 1 and 9 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.