Learn More
Medchemexpress LLC Mavorixafor trihydrochloride | 2309699-17-8 | 99.8% | C21H30Cl3N5 | 1 ML
SDP
Catalog No. 5000357549
Encompass_Preferred
Mavorixafor trihydrochloride (AMD-070 trihydrochloride) is a potent, selective, and orally available CXCR4 antagonist with an IC50 value of 13 nM against CXCR4 125I-SDF binding. It also inhibits the replication of T-tropic HIV-1 (NL4.3 strain) in MT-4 cells and PBMCs with an IC50 of 1 nM and 9 nM, respectively. This compound can be used for the study of WHIM syndrome.
  • Potent, selective, and orally available CXCR4 antagonist
  • Inhibits replication of T-tropic HIV-1
  • Useful for the study of WHIM syndrome
Catalog No. 50-003-57549 Supplier Medchemexpress LLC Supplier No. HY50101A10MM/1ML
Mavorixafor trihydrochloride (AMD-070 trihydrochloride) is a potent, selective, and orally available CXCR4 antagonist with an IC50 value of 13 nM against CXCR4 125I-SDF binding. It also inhibits the replication of T-tropic HIV-1 (NL4.3 strain) in MT-4 cells and PBMCs with an IC50 of 1 nM and 9 nM, respectively. This compound can be used for the study of WHIM syndrome.
  • Potent, selective, and orally available CXCR4 antagonist
  • Inhibits replication of T-tropic HIV-1
  • Useful for the study of WHIM syndrome