Learn More
Medchemexpress LLC Mavorixafor trihydrochloride | 2309699-17-8 | 99.8% | C21H30Cl3N5 | 2 MG
SDP
Catalog No. 5000358102
Encompass_Preferred
Mavorixafor trihydrochloride is a potent, selective, and orally available CXCR4 antagonist. It inhibits the replication of T-tropic HIV-1 and can be used for the study of WHIM syndrome.
  • Potent, selective, and orally available CXCR4 antagonist
  • Inhibits replication of T-tropic HIV-1 (NL4.3 strain) in MT-4 cells with an IC50 of 1 nM
  • Inhibits replication of T-tropic HIV-1 (NL4.3 strain) in PBMCs with an IC50 of 9 nM
  • IC50 value of 13 nM against CXCR4 125I-SDF binding
  • Used for the study of WHIM syndrome
Catalog No. 50-003-58102 Supplier Medchemexpress LLC Supplier No. HY50101A2MG
Mavorixafor trihydrochloride is a potent, selective, and orally available CXCR4 antagonist. It inhibits the replication of T-tropic HIV-1 and can be used for the study of WHIM syndrome.
  • Potent, selective, and orally available CXCR4 antagonist
  • Inhibits replication of T-tropic HIV-1 (NL4.3 strain) in MT-4 cells with an IC50 of 1 nM
  • Inhibits replication of T-tropic HIV-1 (NL4.3 strain) in PBMCs with an IC50 of 9 nM
  • IC50 value of 13 nM against CXCR4 125I-SDF binding
  • Used for the study of WHIM syndrome