Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MAVS protein, human (His-SUMO) | >85.0% | 50UG
SDP

Catalog No. 5000174625
Encompass_Preferred

MAVS Protein, Human (His-SUMO) is a recombinant human mitochondrial antiviral-signaling protein expressed in Escherichia coli and supplied as a lyophilized powder with N-terminal 6xHis and SUMO tags. It is provided for research use and formulated to support stability and downstream assays.

  • Recombinant human protein produced in E. coli for consistent sequence and yield.
  • N-terminal 6xHis and SUMO tags facilitate purification and improve solubility.
  • Lyophilized formulation with trehalose for improved storage stability.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive applications.
  • Purity >85% by reducing SDS-PAGE for reproducible experimental results.
  • Store at -20°C; after reconstitution, aliquot and store at -20°C or -80°C for long term.

Catalog No. 50-001-74625 Supplier Medchemexpress LLC Supplier No. HYP70308250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MAVS Protein, Human (His-SUMO) is a recombinant human mitochondrial antiviral-signaling protein expressed in Escherichia coli and supplied as a lyophilized powder with N-terminal 6xHis and SUMO tags. It is provided for research use and formulated to support stability and downstream assays.

  • Recombinant human protein produced in E. coli for consistent sequence and yield.
  • N-terminal 6xHis and SUMO tags facilitate purification and improve solubility.
  • Lyophilized formulation with trehalose for improved storage stability.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive applications.
  • Purity >85% by reducing SDS-PAGE for reproducible experimental results.
  • Store at -20°C; after reconstitution, aliquot and store at -20°C or -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.