Learn More
Cayman Chemical MAVS MoncLNal ANTIBDYClo 100ug
SDP
Catalog No. 502934001
Encompass_Preferred
Immunogen: full-length recombinant human MAVS protein • Host: Mouse • Species Reactivity: Human, mouse • Cross Reactivity: N/A • Applications: ELISA, IHC, and WB • MW: 56.5 kDa from human samples
Catalog No. 50-293-4001 Supplier Cayman Chemical Supplier No. 26355100 UG
Immunogen: full-length recombinant human MAVS protein • Host: Mouse • Species Reactivity: Human, mouse • Cross Reactivity: N/A • Applications: ELISA, IHC, and WB • MW: 56.5 kDa from human samples