Learn More
Medchemexpress LLC MAX-40279 hemifumarate | 2388506-43-0 | ≥98% | C26H27FN6O5S | 50 MG
SDP
Catalog No. 5000353283
Encompass_Preferred
MAX-40279 hemifumarate is a dual and potent inhibitor of FLT3 kinase and FGFR kinase. It has the potential for the research of acute myelogenous leukemia (AML).
  • Dual and potent inhibitor of FLT3 kinase and FGFR kinase.
  • Potential for research of acute myelogenous leukemia (AML).
Catalog No. 50-003-53283 Supplier Medchemexpress LLC Supplier No. HY145723B50MG
MAX-40279 hemifumarate is a dual and potent inhibitor of FLT3 kinase and FGFR kinase. It has the potential for the research of acute myelogenous leukemia (AML).
  • Dual and potent inhibitor of FLT3 kinase and FGFR kinase.
  • Potential for research of acute myelogenous leukemia (AML).