Learn More
Medchemexpress LLC MAX-40279 hemifumarate | 2388506-43-0 | 99.6% | C22H23FN6OS·1/2C4H4O4 | 10MG
SDP
Catalog No. 5000258507
Encompass_Preferred
MAX-40279 hemifumarate is a small-molecule research inhibitor that targets FLT3 and FGFR kinases. It is orally active and retains activity against clinically relevant FLT3 mutants (for example, D835Y), making it suitable for studies of kinase inhibitor resistance and acute myelogenous leukemia (AML) signaling. It also inhibits NDRG1 phosphorylation at Ser330 and suppresses endothelial-to-mesenchymal transition (EndMT).
  • Dual FLT3 and FGFR kinase inhibition
  • Orally active and effective against FLT3 mutants such as D835Y
  • Useful for AML research and studies of inhibitor resistance
  • Inhibits NDRG1 phosphorylation at Ser330 and suppresses EndMT
  • High purity (~99.6%) and fully characterized chemical properties
Catalog No. 50-002-58507 Supplier Medchemexpress LLC Supplier No. HY145723B10MG
MAX-40279 hemifumarate is a small-molecule research inhibitor that targets FLT3 and FGFR kinases. It is orally active and retains activity against clinically relevant FLT3 mutants (for example, D835Y), making it suitable for studies of kinase inhibitor resistance and acute myelogenous leukemia (AML) signaling. It also inhibits NDRG1 phosphorylation at Ser330 and suppresses endothelial-to-mesenchymal transition (EndMT).
  • Dual FLT3 and FGFR kinase inhibition
  • Orally active and effective against FLT3 mutants such as D835Y
  • Useful for AML research and studies of inhibitor resistance
  • Inhibits NDRG1 phosphorylation at Ser330 and suppresses EndMT
  • High purity (~99.6%) and fully characterized chemical properties