Learn More
Aviva Systems Biology Corp MAX Antibody - middle region (ARP38145_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38145_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MAX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAX. Alternate names: orf1, bHLHd4, bHLHd5, bHLHd6, bHLHd7, bHLHd8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56535