Learn More
Aviva Systems Biology Corp MAX Antibody - N-terminal region (ARP39844_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39844_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human MAX. Alternate names: MGC10775, MGC11225, MGC18164, MGC34679, MGC36767, orf1, bHLHd4, bHLHd5, bHLHd6, bHLHd7, bHLHd8ApplicationWB. Species reactivity: Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-014-53156