Learn More
BD MAX™ Cdiff
Catalog No. B443418 Shop All Waters Corporation Products
Change view
Click to view available options
Detectable Analytes:
C. difficile Toxin B
Catalog No. Detectable Analytes
B443418 C. difficile Toxin B
1 options
This item is not returnable. View return policy
Catalog No. B443418 Supplier BD Supplier No. 443418
Add to Cart
Add to Cart
This item is not returnable. View return policy
BD MAX™ Cdiff is a test for patients suspected of having CDI through the detection of the C. difficile toxin B gene (tcdB) from liquid or soft stool.
The BD MAX™ Cdiff assay performed on the BD MAX System is an automated in vitro diagnostic test for the direct, qualitative detection of the Clostridium difficile toxin B gene (tcdB) in human liquid or soft stool specimens from patients suspected of having Clostridium difficile infection (CDI). The test, performed directly on the specimen, utilizes real-time polymerase chain reaction (PCR) for the amplification of Clostridium difficile toxin B gene DNA and fluorogenic target-specific hybridization probes for the detection of the amplified DNA. The BD MAX Cdiff assay is intended to aid in the diagnosis of CDI.
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes C. difficile Toxin B
Sample Type human liquid or soft stool specimens
Includes BD MAX™ Cdiff Extraction Tubes (24), BD MAX™ Cdiff Sample Buffer Tubes (24), BD MAX™ Septum Caps (25), BD MAX™ Cdiff Reagent Strips (24), BD MAX™ Cdiff Master Mix (24)