Learn More
Fine Science Tools MAX DETAIL SPECTACLES-2X
SDP
Catalog No. NC0263902
Encompass
Product: Max Details Spectacles/ Weight: 49g/ Magnification: 2x/ Working Distance: 40cm/ Features: Adjustable focal length for individual eye/ Unit: Each
This item is not returnable. View return policy
Catalog No. NC0263902 Supplier Fine Science Tools Supplier No. 2803020
Product: Max Details Spectacles/ Weight: 49g/ Magnification: 2x/ Working Distance: 40cm/ Features: Adjustable focal length for individual eye/ Unit: Each