Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD MAX™ DNA MMK

Catalog No. B442848 Shop All Waters Corporation Products
Change view
Click to view available options
For Use With (Equipment):
BD MAX™ System, BD MAX™ ExK DNA (1-4)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
B442848 BD MAX™ System, BD MAX™ ExK DNA (1-4)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B442848 Supplier BD Supplier No. 442848
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A complete PCR reagent mix for amplification of DNA.

BD MAX™ DNA MMK is a dried down PCR reagent mix containing dNTPs, MgCl2, Hot Start DNA polymerase, buffers.
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
For Use With (Equipment) BD MAX™ System, BD MAX™ ExK DNA (1-4)
Includes BD MAX™ DNA MMK Master Mix (24), DNA Primers and Probes Diluent (0.6 mL)
Product Type dried down PCR reagent mix containing dNTPs, MgCl2, Hot Start DNA polymerase, buffers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.