Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD MAX™ ExK™ DNA-1

Catalog No. B442817 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Sample Type:
Plasma, Serum, Neat Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
B442817 Plasma, Serum, Neat Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B442817 Supplier BD Supplier No. 442817
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A kit for extracting DNA from plasma, serum, or neat urine specimens on the BD MAX™ System.

The BD MAX™ ExK™ DNA-1 is intended to extract DNA from plasma, serum or neat urine specimens. The DNA Extraction Tube contains a Sample Processing Control sequence from Drosophila melanogaster (GenBank: AC246436; nucleotides 35779 to 35978) cloned in a pUC119 derivative (GenBank: U07650.1). The BD MAX ExK DNA-1 has not been validated for use with any specific test method.
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Sample Type Plasma, Serum, Neat Urine
For Use With (Equipment) BD MAX™ System
For Use With (Application) DNA Extraction
Includes BD MAX™ ExK™ DNA Extraction Tubes (24), BD MAX™ ExK™ DNA-1 Sample Buffer Tubes (24), BD MAX™ Septum Caps (25), BD MAX™ ExK™ DNA-1 Reagent Strips (24), BD MAX™ Conical Tubes (24)
Product Type DNA Extraction Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.