Learn More
Description
Specifications
Specifications
| Format | Kit |
| Sample Type | Cerebrospinal Fluid, Swab (Dry) |
| For Use With (Equipment) | BD MAX™ System |
| For Use With (Application) | DNA Extraction |
| Includes | BD MAX™ ExK™ DNA Extraction Tubes (24), BD MAX™ ExK™ DNA-2 Sample Buffer Tubes (24), BD MAX™ Septum Caps (25), BD MAX™ ExK™ DNA-2 Reagent Strips (24), BD MAX™ Conical Tubes (24) |
| Product Type | Extraction Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.