Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD MAX™ ExK™ TNA-3

Catalog No. B442827 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Sample Type:
Swab Specimen in Universal Transport Media (UTM)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
B442827 Swab Specimen in Universal Transport Media (UTM)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B442827 Supplier BD Supplier No. 442827
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A kit for extracting total nucleic acid (TNA) from swab specimens in Universal Transport Medium (UTM) on the BD MAX™ System.

The BD MAX™ ExK™ TNA-3 is intended to extract total nucleic acid (TNA) from swab specimens in Universal Transport Medium (UTM). The TNA Extraction Tube contains a Specimen Processing Control sequence from a proprietary Armored RNA Quant™ (Asuragen, Inc.).* The BD MAX ExK TNA-3 has not been validated for use with any specific test method.
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Sample Type Swab Specimen in Universal Transport Media (UTM)
For Use With (Equipment) BD MAX™ System
For Use With (Application) Total nucleic acid (TNA) Extraction
Includes BD MAX™ ExK™ TNA Extraction Tubes (24), BD MAX™ ExK™ TNA-3 Sample Buffer Tubes (24), BD MAX™ Septum Caps (25), BD MAX™ ExK™ TNA-3 Reagent Strips (24), BD MAX™ Conical Tubes (24)
Product Type Extraction Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.