Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD MAX™ GBS

Catalog No. B441772 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Sensitivity:
95%
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sensitivity
B441772 95%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B441772 Supplier BD Supplier No. 441772
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BD MAX™ GBS is a fully-automated, real-time PCR method for the detection of GBS from enriched culture. It simplifies detection of GBS colonization in antepartum pregnant women, and rapidly identifies Group B Streptococcus after an overnight Lim Broth enrichment.

The BD MAX™ GBS assay as implemented on the BD MAX System is a qualitative in vitro diagnostic test designed to detect Group B Streptococcus (GBS) DNA in Lim Broth cultures, after incubation for greater than or equal to (≥)18 hours, obtained from vaginal-rectal swab specimens from antepartum pregnant women. The test incorporates automated DNA extraction to isolate the target nucleic acid from the specimen and real-time polymerase chain reaction (PCR) to detect a 124 bp region of the cfb gene sequence of the Streptococcus agalactiae chromosome. Results from the BD MAX GBS assay can be used as an aid in determining colonization status in antepartum women.
TRUSTED_SUSTAINABILITY

Specifications

Sensitivity 95%
Type Group B Streptococcus Testing
Includes BD MAX™ GBS Extraction Tubes (24), BD MAX™ GBS Sample Buffer Tubes (24), BD MAX™ Septum Caps (25), BD MAX™ GBS Reagent Strips (24), BD MAX™ GBS Master Mix (24)
Specificity 0.967
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.