Learn More
Microsolv Technology Corporation Max Rec 1.8Ml Amber Vial 100Pk
Catalog No. NC1836981
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 1.8ml, Maximum Recovery, Screw Top, Amber RSA™ Autosampler Vials and 100 EA matching Screw Caps with Clear AQR Silicone Rubber / Clear PTFE, ultra-pure Septa, fitted in the Light Blue Caps. RSA Brand Easy Purchase Pack. 1/PK.
Catalog No. NC1836981 Supplier Microsolv Technology Corporation Supplier No. 9512S1APRS
Vial & Cap Kit Includes 100 EA 1.8ml, Maximum Recovery, Screw Top, Amber RSA™ Autosampler Vials and 100 EA matching Screw Caps with Clear AQR Silicone Rubber / Clear PTFE, ultra-pure Septa, fitted in the Light Blue Caps. RSA Brand Easy Purchase Pack. 1/PK.