Learn More
Thermco Products Inc MAX REG THRMTR, CERT, TRCBLE
Catalog No. NC0556533
Encompass_Preferred
NIST compliant, factory certified glass Hg thermometer, 80 deg./135 deg.C, 0.5C div, TOTAL imm, w/safety coat, certified at 121 deg.C, 200mm
Catalog No. NC0556533 Supplier Thermco Products Inc Supplier No. ACC80135SCFC
NIST compliant, factory certified glass Hg thermometer, 80 deg./135 deg.C, 0.5C div, TOTAL imm, w/safety coat, certified at 121 deg.C, 200mm