Learn More
BD MAX™ Respiratory Viral Panel (RVP)
Catalog No. B445373 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Detectable Analytes:
SARS-CoV-2, Influenza A and B, Respiratory Syncytial Virus (RSV)
Catalog No. Detectable Analytes
B445373 SARS-CoV-2, Influenza A and B, Respiratory Syncytial Virus (RSV)
1 options
This item is not returnable. View return policy
Catalog No. B445373 Supplier BD Supplier No. 445373
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
This Respiratory Viral Panel is able to detect and differentiate SARS-CoV2, influenza A, influenza B, and RSV.
BD Respiratory Viral Panel for BD MAXTM System, you can simultaneously detect and differentiate SARS-CoV-2, influenza A, influenza B, and/or RSV from one sample in a single run. The test uses a single nasal swab or a single nasopharyngeal swab sample to determine if a patient has COVID-19 or the flu or RSV. 510K Approved.
  • Simultaneously detect and differentiate SARS-CoV-2, influenza A, influenza B, and/or RSV from one sample in a single run
  • Single nasal swab or a single nasopharyngeal swab sample is used for testing
  • 510K approved
TRUSTED_SUSTAINABILITY
Specifications
CE Marker CE-IVD
Clia Complexity High, Moderate
Detectable Analytes SARS-CoV-2, Influenza A and B, Respiratory Syncytial Virus (RSV)
Disposable Single-use
Sterility Non-sterile
Form Ready-to-Use (RTU)