Learn More
Description
- With this RVP system, you can simultaneously detect and differentiate SARS-CoV-2, influenza A, influenza B, and/or RSV from one sample in a single run
- The test uses a single nasal swab or a single nasopharyngeal swab sample to determine if a patient has COVID-19 or the flu or RSV
Specifications
Specifications
| CE Marker | Yes |
| Clia Complexity | Moderate |
| Detectable Analytes | SARS-CoV-2, Influenza A and B, Respiratory Syncytial Virus (RSV) |
| Detection Method | Single nasal swab or a single nasopharyngeal swab |
| Format | Kit |
| Sample Type | Nasal or Nasopharyngeal |
| Sensitivity | High |
| Type | PCR Assay |
| Disposable | Reusable |
| Sterility | Non-sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.