Learn More
Metro Max Stationary Security Shelving Unit
Catalog No. 01001881 Shop All Metro Products
Change view
Click to view available options
Dimensions (L x W x H):
28.1875 x 50.875 x 66.1875 in.
28.1875 x 62.875 x 66.1875 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
01-001-881 28.1875 x 50.875 x 66.1875 in.
01-000-998 28.1875 x 62.875 x 66.1875 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-001-881 Supplier Metro Supplier No. MQSEC55ESL
Add to Cart
Add to Cart
MetroMax Stationary Security Shelving Unit with SlideSecure Sliding Door.
  • Keep sensitive items safe from loss or pilferage.
  • Heavy-gauge open wire construction keeps contents visible at all times, making it easy to check inventory.
  • Metro SlideSecure Space Saving Doors provide the same level of protection as traditional security units with the space saving convenience of our sliding door system.
  • Units include two shelves which must be used as the top and bottom shelves of the unit.
  • Add optional intermediate shelves for increased storage capabilities.
  • Compatible with all MetroMax i, 4 and Q shelves.
  • Padlock sold separately.
Specifications
Product Type Rack
Color Gray
Material Epoxy-coated Steel
Autoclavable Not Autoclavable
Dimensions (L x W x H) 28.1875 x 50.875 x 66.1875 in.
With Casters No