Learn More
BD Vaginal Panel
Catalog No. B443712 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Detectable Analytes:
Bacterial Vaginosis, Vulvovaginal Candidiasis, Trichomonas Vaginalis
Catalog No. Detectable Analytes
B443712 Bacterial Vaginosis, Vulvovaginal Candidiasis, Trichomonas Vaginalis
1 options
This item is not returnable. View return policy
Catalog No. B443712 Supplier BD Supplier No. 443712
Add to Cart
Add to Cart
This item is not returnable. View return policy
The BD Vaginal Panel is the most-comprehensive and first FDA market-authorized, microbiome-based, polymerase chain reaction (PCR) assay that directly detects the 3 most common infectious causes of vaginitis - Bacterial vaginosis, vulvovaginal candidiasis and trichomoniasis.
The BD Vaginal Panel performed on the BD MAX™ System is an automated qualitative in vitro diagnositc test for the direct detection of DNA targets from bacteria associated with bacterial vaginosis (qualitative results reported based on detection and quantitation of targeted organism markers), Candida species associated with vulvovaginal candidiasis, and Trichomonas vaginalis from vaginal swabs in patients who are symptomatic for vaginities/vaginosis. The test utilizes real-time polymerase chain reaction (PCR) for the amplification of specific DNA targets and utilizes fluorogenic target-specific hybridization probes to detect and differentiate DNA from: Bacterial vaginosis markers (Individual markers not reported) Lactobacillus spp. (L. crispatus and L. jensenii) Gardnerella vaginalis, Atopobium vaginae, Bacterial Vaginosis Associated Bacteria-2 (BVAB-2), Megasphaera-1, Candida spp. (C. albicans, C. tropicalis, C. parapsilosis, C. dubliniensis), C. glabrata, C. krusei, T. vaginalis. The BD Vaginal Panel is intended to aid in the diagnosis of vaginal infections in women with a clinical presentation consistent with bacterial vaginosis, vulvovaginal candidiasis, and trichomoniasis.
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Bacterial Vaginosis, Vulvovaginal Candidiasis, Trichomonas Vaginalis
Type Microbiome-based, PCR Assay 
Includes BD Vaginal Panel Extraction Tubes (24), BD Septum Caps (25), BD Vaginal Panel Reagent Strips (24), BD Vaginal Panel Vaginitis Master Mix (24), BD Vaginal Panel Vaginosis Master Mix (24)