Learn More
Medchemexpress LLC HY-32339 1mg Medchemexpress, Maxacalcitol CAS:103909-75-7 Purity:>98%
SDP
Catalog No. 502029308
Encompass_Preferred
Medchemexpress, HY-32339 1mg Maxacalcitol CAS:103909-75-7 Maxacalcitol (22-Oxacalcitriol) is non-calcemic vitamin D3 analog and ligand of VDR-like receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9308 Supplier Medchemexpress LLC Supplier No. HY323391MG
Medchemexpress, HY-32339 1mg Maxacalcitol CAS:103909-75-7 Maxacalcitol (22-Oxacalcitriol) is non-calcemic vitamin D3 analog and ligand of VDR-like receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.