Learn More
Medchemexpress LLC Maxacalcitol-d6 | 2070009-38-8 | 99.6% | 424.65 | C26H36D6O4 | 10 MG
SDP
Catalog No. 5000285990
Encompass_Preferred
Maxacalcitol-d6 is the deuterated form of maxacalcitol (22-oxacalcitriol), a non-calcemic vitamin D3 analog and vitamin D receptor (VDR) ligand used as a labeled reference standard for research. It is supplied as a high-purity solid (99.63%) in a 10 mg pack and is intended for analytical applications including mass spectrometry, pharmacokinetic studies, and VDR pathway research.
  • Deuterated internal standard for quantitative mass spectrometry.
  • Non-calcemic vitamin D3 analog and VDR ligand.
  • High purity (99.63%) suitable for analytical and bioanalytical assays.
  • Solid white to off-white form with good stability under recommended storage.
  • Supplied in 10 mg packs for laboratory use.
Catalog No. 50-002-85990 Supplier Medchemexpress LLC Supplier No. HY1532910MG
Maxacalcitol-d6 is the deuterated form of maxacalcitol (22-oxacalcitriol), a non-calcemic vitamin D3 analog and vitamin D receptor (VDR) ligand used as a labeled reference standard for research. It is supplied as a high-purity solid (99.63%) in a 10 mg pack and is intended for analytical applications including mass spectrometry, pharmacokinetic studies, and VDR pathway research.
  • Deuterated internal standard for quantitative mass spectrometry.
  • Non-calcemic vitamin D3 analog and VDR ligand.
  • High purity (99.63%) suitable for analytical and bioanalytical assays.
  • Solid white to off-white form with good stability under recommended storage.
  • Supplied in 10 mg packs for laboratory use.