Learn More
Medchemexpress LLC Maxacalcitol-d6 | 2070009-38-8 | C26H36D6O4 | 5 MG
SDP
Catalog No. 5000372543
Encompass_Preferred
Maxacalcitol-d6 is the deuterated form of Maxacalcitol (22-Oxacalcitriol), a non-calcemic vitamin D3 analog and a VDR ligand of VDR-like receptors. This compound is intended for research use only and is not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-72543 Supplier Medchemexpress LLC Supplier No. HY153295MG
Maxacalcitol-d6 is the deuterated form of Maxacalcitol (22-Oxacalcitriol), a non-calcemic vitamin D3 analog and a VDR ligand of VDR-like receptors. This compound is intended for research use only and is not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS