Learn More
Bachem Maxadilan Trifluoroacetate
SDP
Supplier:  Bachem 4067317.0500
Encompass_Preferred
Maxadilan Trifluoroacetate, H-Cys-Asp-Ala-Thr-Cys-Gln-Phe-Arg-Lys-Ala-Ile-Asp-Asp-Cys-Gln-Lys-Gln-Ala-His-His-Ser-Asn-Val-Leu-Gln-Thr-Ser-Val-Gln-Thr-Thr-Ala-Thr-Phe-Thr-Ser-Met-Asp-Thr-Ser-Gln-Leu-Pro-Gly-Asn-Ser-Val-Phe-Lys-Glu-Cys-Met-Lys-Gln-Lys-Lys-Lys-Glu-Phe-Lys-Ala-NH2, 135374-80-0, C291H466N86O94S6, Mr 6865.82, 0.5mg. Maxadilan, isolated from the salivary gland of the sand fly Lutzomyia longipalpis, is a potent vasodilator. The peptide specifically and potently activates the mammalian PAC1 receptor, one of the three receptors for PACAP.
Catalog No. 50-260-121