Learn More
IBI SCIENTIFIC 3P Maxi Flex Tube Kit, 6 Kda MWCO, 100/pk
Catalog No. NC0753528
Encompass_Preferred
MAXI (6K MWCO) FlexTube Kit - 100/Pack IBI MAXI FlexTubes combine two modes of action, electro-elution of macromolecules from polyacrylamide or agarose gels and dialysis or buffer exchange of samples ranging in size from 0.1-3ml.
Catalog No. NC0753528 Supplier IBI SCIENTIFIC 3P Supplier No. IB48235
MAXI (6K MWCO) FlexTube Kit - 100/Pack IBI MAXI FlexTubes combine two modes of action, electro-elution of macromolecules from polyacrylamide or agarose gels and dialysis or buffer exchange of samples ranging in size from 0.1-3ml.