Learn More
IBI SCIENTIFIC 3P Maxi Flextube Kit, 3.5KDa, 50/pk
Catalog No. NC1657854
Encompass_Preferred
IBI MAXI FlexTubes are typically used for the following applications: dialysis, gel extraction, electro-elution, and de-salting. The design of the FlexTube allows for easy loading and unloading with a standard pipette tip and screw-cap top. Multiple tubes can be placed in one buffer tank for high-throughput applications with minimal hands-on time.
Catalog No. NC1657854 Supplier IBI SCIENTIFIC 3P Supplier No. IB48210
IBI MAXI FlexTubes are typically used for the following applications: dialysis, gel extraction, electro-elution, and de-salting. The design of the FlexTube allows for easy loading and unloading with a standard pipette tip and screw-cap top. Multiple tubes can be placed in one buffer tank for high-throughput applications with minimal hands-on time.