Learn More
Hoefer, Inc Hoefer™ SUB25 Maxi-Plus Standard Submarine Gel Electrophoresis Unit
SDP
Catalog No. NC2741108
Encompass_Preferred
Ideal for high throughput screening of nucleic acids, particularly PCR products and in genotyping studiesFeatures & Benefits:Large format—25 x 30 cm gel tray is ideal for high throughput techniquesMultichannel pipette compatible combs—with a maximum 52 sample throughput-reduce gel loading timeBuffer recirculation ports—may be connected to a peristaltic pump for buffer recirculation during electrophoresis to maintain buffer pH and prevent ionic gradient formationUV-transparent running traySide handles—for safe and easy transportationIncludes:Buffer Chamber Assembly; Safety Lid; High Voltage Leads; Running Tray w/Casting Gates; 26 well combs, 6 pcs; Colored Loading Strips; Buffer Recirculation Ports, 2 pcs;Specifications:Gel Dimensions (W x L): 25 x 30 cmMaximum Buffer Volume: 3 LMaximum Power Settings: 1000 V, 500 mA, 500 WMaximum Sample Capacity: 624Unit Dimensions (W x L x H): 35 x 56 x 9 cm
Catalog No. NC2741108 Supplier Hoefer, Inc Supplier No. SUB25
Ideal for high throughput screening of nucleic acids, particularly PCR products and in genotyping studiesFeatures & Benefits:Large format—25 x 30 cm gel tray is ideal for high throughput techniquesMultichannel pipette compatible combs—with a maximum 52 sample throughput-reduce gel loading timeBuffer recirculation ports—may be connected to a peristaltic pump for buffer recirculation during electrophoresis to maintain buffer pH and prevent ionic gradient formationUV-transparent running traySide handles—for safe and easy transportationIncludes:Buffer Chamber Assembly; Safety Lid; High Voltage Leads; Running Tray w/Casting Gates; 26 well combs, 6 pcs; Colored Loading Strips; Buffer Recirculation Ports, 2 pcs;Specifications:Gel Dimensions (W x L): 25 x 30 cmMaximum Buffer Volume: 3 LMaximum Power Settings: 1000 V, 500 mA, 500 WMaximum Sample Capacity: 624Unit Dimensions (W x L x H): 35 x 56 x 9 cm