Learn More
LUNA NANOTECH INC Maxi Silica Spin Columns, 10 Columns
Supplier:  LUNA NANOTECH INC USP00810N
Encompass_Preferred
PuroSPIN™ MAXI Spin Columns for DNA / RNA Extraction and Purification, 10 columns
Have you ever ran out of columns that come with your favorite plasmid extraction kit but still had a lot of left-over buffer reagents remaining? Instead of buying a new expensive kit you can use PuroSPIN™ columns with the left-over buffers as a cost-effective alternative. Simply use PuroSPIN™ columns while following the manufacturer-specific protocols.
Nucleic acid binding capacity: 2.3 mg of genomic DNA, 500 µg of plasmid DNA
Compatible as cost-effective replacement columns for Plasmid DNA purification kits from Qiagen, ThermoFisher, Bio-Rad, Promega and other manufacturers
PuroSPIN™ MAXI silica spin columns are designed for fast, simple and efficient extraction of Plasmid DNA from bacterial cultures
Catalog No. 50-256-6903