Learn More
Shockwatch Div Mri MaxiLog Alert -80C Single Use
SDP
Catalog No. NC2598420
Encompass_Preferred
A single-use, pre-programmed electronic temperature data recorder for measuring and documenting temperatures of dry ice shipments including vaccines, biological material, chemicals, pharmaceuticals, and other temperature sensitive products.•-80°C to 30°C / -113°F to 86°F•8K Memory (approx. 8,000 data points)•1-year operating life•8.8 cm x 3.49 cm x 1.58 cm / 3.5” x 1.38” x 0.62”•3 oz / 65g•±0.5°C at 0 to 10°C / ±0.9°C at extremes •±0.9°F at 32° to 50° F / ±3.6°F at extremes•0.1°C temperature resolution•1 year warranty•ER2045T Lithium coin cell •10 per box
Catalog No. NC2598420 Supplier Shockwatch Div Mri Supplier No. 8114S
A single-use, pre-programmed electronic temperature data recorder for measuring and documenting temperatures of dry ice shipments including vaccines, biological material, chemicals, pharmaceuticals, and other temperature sensitive products.•-80°C to 30°C / -113°F to 86°F•8K Memory (approx. 8,000 data points)•1-year operating life•8.8 cm x 3.49 cm x 1.58 cm / 3.5” x 1.38” x 0.62”•3 oz / 65g•±0.5°C at 0 to 10°C / ±0.9°C at extremes •±0.9°F at 32° to 50° F / ±3.6°F at extremes•0.1°C temperature resolution•1 year warranty•ER2045T Lithium coin cell •10 per box