Learn More
Shockwatch Div Mri MaxiLog-PDF V3
SDP
Catalog No. NC2598429
Encompass_Preferred
A single-use, 90-day temperature data logger with USB connectivity and PDF reports.•-30°C to 70°C / -22°F to 158°F•±0.3°C / ±0.54°F •0.1°C temperature resolution•14K Memory •1-year operating life•94 mm x 20 mm x 7 mm / 3.7” x 0.8” x 0.27”•0.37 oz / 10g•1 year warranty•CR2032 Lithium coin cell •10 per box
Catalog No. NC2598429 Supplier Shockwatch Div Mri Supplier No. 7050
A single-use, 90-day temperature data logger with USB connectivity and PDF reports.•-30°C to 70°C / -22°F to 158°F•±0.3°C / ±0.54°F •0.1°C temperature resolution•14K Memory •1-year operating life•94 mm x 20 mm x 7 mm / 3.7” x 0.8” x 0.27”•0.37 oz / 10g•1 year warranty•CR2032 Lithium coin cell •10 per box