Learn More
3M™ Maxim™ 2X2 Safety Goggles
Catalog No. 190600424 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Gray
No. per Case:
10/Cs.
Catalog No. Lens Tint No. per Case
19-060-0424 Gray 10/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 19-060-0424 Supplier 3M™ Supplier No. 7000127566
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Versatile design for tacticle use
Lightweight safety goggle was designed to help protect from sun, wind, dust and flying debris.
  • Dual-lens design enhances sightline and field of vision
  • Vented air seal designed to help minimize fogging
  • Featuring flexible fit temples
  • Interchangeable temples and broad elastic strap
  • 3M Catalog# 40686-00000-10 , 3M Catalog# 40687-00000-10
Compliance
Meets the High Impact Requirements of ANSI Z87.1-2003.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2003
Lens Tint Gray
Product Type Safety goggles
For Use With (Application) Provide protection against hazards such as dust and chemical splash
Frame Color Black
No. per Case 10/Cs.