Learn More
3M™ Maxim™ Eyewear
Catalog No. 19121575 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Temple Style:
Adjustable
Catalog No. Lens Tint Temple Style
19-121-575 Clear Adjustable
1 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-121-575 Supplier 3M™ Supplier No. 7010045586
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Contemporary, athletic styling
Feauture polycarbonate lenses that absorb 99.9% UV.
  • Three-position pantascopic lens angle adjustment
  • Dual-injected cushioned browguard
  • Soft nose adjustable nose bridge
  • Adjustable temple lengths
  • Ventilation channel helps to prevent fogging
  • 3M Catalog# 13250-00000-20
Compliance
Meets ANSI Z87.1-2015 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Clear
Frame Color Black
Model 13250-00000-20
Temple Style Adjustable
Lens Material Polycarbonate