Learn More
Fisherbrand™ Maxima C Plus Gasket Sets
Catalog No. p-159770 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With:
M2C, M4C Vacuum Pumps
M8C, M12C Vacuum Pumps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
01-257-3B M8C, M12C Vacuum Pumps
01-257-3A M2C, M4C Vacuum Pumps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-257-3B
Only null left
Add to Cart
Add to Cart
Replacement gasket sets for use with Maxima C vacuum pumps
Contain all the Viton™ O-rings, shaft seals, and gaskets needed to rebuild the pump.
†For M4C, M6C or M8C vacuum pumps designed before September 1999 (S/N before FKK090000).
Specifications
For Use With M8C, M12C Vacuum Pumps
Includes Viton™ O-rings, shaft seals and gaskets
Type Gasket Set