Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ MaximaDry™ Diaphragm Vacuum Pump

Catalog No. 1388014 Shop All Fisherbrand Products
Change view
Click to view available options
Flow Rate:
20 L/min.
35 L/min.
40 L/min.
Max. Vacuum:
1.5 torr
6 torr
65 torr
75 torr
Voltage:
115 V
230 V
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Max. Vacuum Voltage
13-880-14 20 L/min. 75 torr 115 V
13-880-18 20 L/min. 6 torr 115 V
13-880-15 20 L/min. 75 torr 230 V
13-880-22 35 L/min. 1.5 torr 115 V
13-880-20 35 L/min. 6 torr 115 V
13-880-16 40 L/min. 65 torr 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 13-880-14
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Requires

3/8 (0.375) in./0.95 cm I.D. tubing

Clean, reliable, oil-free vacuum pumps are ideal for low to high boiling point rotary evaporation and distillation, filtration, blotting, and solid phase extraction.

Clean, reliable, oil-free vacuum pumps are ideal for high boiling point rotary evaporation and distillion, vacuum concentration, and larger vacuum ovens (>0.5 cu.ft.)

Quiet, reliable operation without the hazards of handling pump oil or contaminated water. Excellent replacement for rotary vane pumps and water aspirators, especially in applications where chemical vapors are encountered.

  • Withstands aggressive gases and vapors — can be used with aggressive solvents without the risk of damaging the pump
  • Single-piece PTFE coated diaphragm and molded PTFE pump heads provide chemical resistance
  • High-flow, multiport valves are made from tough FFPM fluoropolymer to withstand pumping heavy solvent vapors and liquid condensate without a loss in performance
  • Fan-cooled (TEFC) motor is rated for continuous operation and never needs lubrication
  • Compact design fits easily on a benchtop or in a fume hood
  • Setup and disassembly are quick and easy and do not require any special tools
  • Available in either 115 V/60 Hz or 230 V/50 Hz

Plugs into electrical outlet; no hardwiring required.

Specifications

Max. Vacuum 75 torr
Electrical Requirements 115 V 60 Hz
Dimensions (L x W x H) 13 x 6.5 x 8.5 in. (33 x 16.5 x 22 cm)
Model MaximaDry 20 to 75; 115V
Flow Rate 20 L/min.
Frequency 60 Hz
Pump Type Diaphragm Vacuum Pump
Enclosure Rating IP44
Applications Blotting
Maximum Pressure 15 psig
Voltage 115 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.