Learn More
Boca Scientific Inc Maxime PCR PreMix Kit (i-StarTaq™), HotStart, 96 Tubes
Catalog No. NC9761838
Encompass
Maxime PCR PreMix Kit (i-StarTaq™), HotStart contains all components necessary for a PCR reaction in a dried and stable format in each tube. PCR can be carried out immediately with the addition of the template, primer and distilled water, reducing the risk of pipetting error. Gel loading electrophoresis is also possible as the tubes contain loading dye. This kit features Intron’s i-StarTaq™ DNA Polymerase. This polymerase implements a chemical-based HotStart function that is not antibody-based and improves specificity and accuracy. This kit has excellent stability due to oxidation and moisture prevention achieved by a vacuum compression manufacturing method. Each tube contains i-StarTaq™ DNA Polymerase, dNTPs, reaction buffer(10x), and gel loading buffer.
Catalog No. NC9761838 Supplier Boca Scientific Inc Supplier No. 25165
Maxime PCR PreMix Kit (i-StarTaq™), HotStart contains all components necessary for a PCR reaction in a dried and stable format in each tube. PCR can be carried out immediately with the addition of the template, primer and distilled water, reducing the risk of pipetting error. Gel loading electrophoresis is also possible as the tubes contain loading dye. This kit features Intron’s i-StarTaq™ DNA Polymerase. This polymerase implements a chemical-based HotStart function that is not antibody-based and improves specificity and accuracy. This kit has excellent stability due to oxidation and moisture prevention achieved by a vacuum compression manufacturing method. Each tube contains i-StarTaq™ DNA Polymerase, dNTPs, reaction buffer(10x), and gel loading buffer.