Learn More
Analytical Sales & Services Maximum Recovery - 1.5mL Amber Tear Drop Screw Vials and NON-BONDED Caps with PTFE/Silicone Liners
Catalog No. 5000805877
Encompass
Advantage™ Unassembled Maximum Recovery Vial Kit\n\nContains:\n• 9mm,12 x 32, 1.5mL Amber Glass Tear Drop Screw Vials\n• NON-BONDED 9mm Screw Caps with PTFE/Silicone Liners
Catalog No. 50-008-05877 Supplier Analytical Sales & Services Supplier No. 99261A
Advantage™ Unassembled Maximum Recovery Vial Kit\n\nContains:\n• 9mm,12 x 32, 1.5mL Amber Glass Tear Drop Screw Vials\n• NON-BONDED 9mm Screw Caps with PTFE/Silicone Liners