Learn More
Analytical Sales & Services Maximum Recovery - 9mm, 12 x 32, 1.2mL Clear Tear Drop Screw Vials and Caps with Pre-Slit, Non Bonded PTFE/Silicone Liners
Catalog No. 5000806288
Encompass
Advantage™ Unassembled Maximum Recovery Vial Kit. Contains: 9mm, 12 x 32mm 1.2mL Clear Glass Tear Drop Screw Vials and 9mm Screw Caps with Pre-Slit Non Bonded PTFE/Silicone Liners
Catalog No. 50-008-06288 Supplier Analytical Sales & Services Supplier No. 86868SL
Advantage™ Unassembled Maximum Recovery Vial Kit. Contains: 9mm, 12 x 32mm 1.2mL Clear Glass Tear Drop Screw Vials and 9mm Screw Caps with Pre-Slit Non Bonded PTFE/Silicone Liners