Learn More
Analytical Sales & Services Maximum Recovery - 9mm, 12 x 32, 1.5mL Amber Tear Drop Screw Vials and Caps with Pre-Slit PTFE/Silicone Liners
Catalog No. 5000805707
Encompass
Advantage™ Unassembled Maximum Recovery Vial Kit. Contains: 9mm, 12 x 32 1.5mL Amber Glass Tear Drop Screw Vials and 9mm Screw Caps with Pre-Slit PTFE/Silicone Liners
Catalog No. 50-008-05707 Supplier Analytical Sales & Services Supplier No. 99290A
Advantage™ Unassembled Maximum Recovery Vial Kit. Contains: 9mm, 12 x 32 1.5mL Amber Glass Tear Drop Screw Vials and 9mm Screw Caps with Pre-Slit PTFE/Silicone Liners