Learn More
Waters Corp Waters Corporation UPLC Method Development Kit, Maximum Selectivity, 4 columns, 3.0 × 50 mm, 1.7 μm
Catalog No. 50791103 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ACQUITY UPLC Method Development Kit offers maximum selectivity for reversed-phase (RP) and HILIC separations. It combines CSH reversed-phase and HILIC stationary phases to retain analytes across a broad range of polarity, ensuring the widest possible separation selectivity.
  • Contains 4 columns per pack.
  • Features 3.0 × 50 mm column dimensions.
  • Includes columns with 1.7 μm CSH and BEH particle sizes.
  • Chemistries include CSH C18, CSH Phenyl-Hexyl, CSH Fluoro-Phenyl, and BEH Amide.
  • Designed for widest separation selectivity.
  • Combines CSH reversed-phase and HILIC stationary phases.
  • Retains analytes encompassing a broad range of polarity.
Catalog No. 50-791-103 Supplier Waters Corp Supplier No. 176002129
Add to Cart
Add to Cart
Request a Quote
This ACQUITY UPLC Method Development Kit offers maximum selectivity for reversed-phase (RP) and HILIC separations. It combines CSH reversed-phase and HILIC stationary phases to retain analytes across a broad range of polarity, ensuring the widest possible separation selectivity.
  • Contains 4 columns per pack.
  • Features 3.0 × 50 mm column dimensions.
  • Includes columns with 1.7 μm CSH and BEH particle sizes.
  • Chemistries include CSH C18, CSH Phenyl-Hexyl, CSH Fluoro-Phenyl, and BEH Amide.
  • Designed for widest separation selectivity.
  • Combines CSH reversed-phase and HILIC stationary phases.
  • Retains analytes encompassing a broad range of polarity.