Learn More
Electron Microscopy Sciences NUNC MaxiSorp™ 96 Well Plates (Polystyrene) Black
SDP
Catalog No. 5031450
Encompass_Preferred
NUNC MaxiSorp plates have been the certified ELISA plate industry standard for years. This same surface now sets a new standard for protein multiplexing. Certified for absorption of IgG; CV of <5% between wells; all results are within 10% from the mean and background of all well within 0.005 absorbance units from the mean Plate Material: Black Polystyrene for protein & antibody multiplexing, fluorescence.
Catalog No. 50-314-50 Supplier Electron Microscopy Sciences Supplier No. 63494BK
Add to Cart
Add to Cart
NUNC MaxiSorp plates have been the certified ELISA plate industry standard for years. This same surface now sets a new standard for protein multiplexing. Certified for absorption of IgG; CV of <5% between wells; all results are within 10% from the mean and background of all well within 0.005 absorbance units from the mean Plate Material: Black Polystyrene for protein & antibody multiplexing, fluorescence.