Learn More
Med Vet International Maxorb Extra Cmc/Alginate Dressing 2 X 2-in, Non-Sterile, 10/Box
Catalog No. 502831436
Encompass_Preferred
Ideal for moderate to heavily draining partial- and full-thickness wounds. Non-sterile Wick fluid vertically to help prevent periwound skin masceration Up to seven day wear time
Catalog No. 50-283-1436 Supplier Med Vet International Supplier No. XMSC7022EPZ
Ideal for moderate to heavily draining partial- and full-thickness wounds. Non-sterile Wick fluid vertically to help prevent periwound skin masceration Up to seven day wear time