Learn More
Active Motif MAXpack™ Immunostaining Media Kit
SDP
Catalog No. NC1841521
Encompass
MAXblock™ Blocking Medium is a protein-based, non-mammalian blocking agent for use in immunofluorescence and immunostaining assays. Superior blocking is achieved utilizing a protein blend that demonstrates no cross-reactivity with secondary antibodies. It has been proven effective at blocking non-specific binding of antibodies in immunofluorescence, as well as in immunohistochemistry.
Catalog No. NC1841521 Supplier Active Motif Supplier No. 15251
MAXblock™ Blocking Medium is a protein-based, non-mammalian blocking agent for use in immunofluorescence and immunostaining assays. Superior blocking is achieved utilizing a protein blend that demonstrates no cross-reactivity with secondary antibodies. It has been proven effective at blocking non-specific binding of antibodies in immunofluorescence, as well as in immunohistochemistry.